Platinum 5% on activated carbon

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Platinium

78-1600-10G 78-1600-50G
100208-962EA 227.88 USD
100208-962 100208-964
Platinum 5% on activated carbon
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 3178
ADR: 4.1 II
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR