Platinum(II) hexafluoroacetylacetonate ≥98% (99.9%-Pt)

Supplier: STREM CHEMICALS INC MS
Warning

78-1550-2G 78-1550-500MG
100196-888EA 1447.94 USD
100196-888 100200-718
Platinum(II) hexafluoroacetylacetonate ≥98% (99.9%-Pt)
Platinum(II) hexafluoroacetylacetonate

Metal Beta-diketonates, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 65 °C (0.1 mmHg) (subl.)
Melting Pt: 143…145 °C
MDL Number: MFCD00058733
CAS Number: 65353-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR