Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum(II)

Supplier: STREM CHEMICALS INC MS
Warning

78-0725-500MG 78-0725-100MG
101419-138EA 985.11 USD
101419-138 101419-136
Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum(II)
Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum(II)
Formula: C₆H₂₁O₃P₃Pt
MW: 429.228 g/mol
MDL Number: MFCD08459361
CAS Number: 173416-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR