(1,5-Cyclooctadiene)dimethylplatinum(II) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Dimethyl(1,5-cyclooctadiene)platinum(II)

78-0700-250MG 78-0700-1G 78-0700-5G
100206-818EA 184 USD
100206-818 100198-038 100210-918
(1,5-Cyclooctadiene)dimethylplatinum(II) ≥99%
(1,5-Cyclooctadiene)dimethylplatinum(II)
Formula: C₁₀H₁₈Pt
MW: 333.33 g/mol
MDL Number: MFCD00064719
CAS Number: 12266-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR