Di-mu-iodobis(ethylenediamine)diplatinum(II) nitrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

78-0600-1G 78-0600-250MG
100211-172EA 795.45 USD
100211-172 100211-170
Di-mu-iodobis(ethylenediamine)diplatinum(II) nitrate ≥99%
Di-mu-iodobis(ethylenediamine)diplatinum(II) nitrate
MDL Number: MFCD00058712
CAS Number: 109998-76-7
UN: 1479
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR