Di-µ-chlorodichlorobis(ethylene)diplatinum(II) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

78-0460-1G 78-0460-5G 78-0460-250MG
100196-376EA 417.54 USD
100196-376 100196-378 100206-816
Di-µ-chlorodichlorobis(ethylene)diplatinum(II) ≥97%
Di-µ-chlorodichlorobis(ethylene)diplatinum(II)
Formula: C₄H₄Cl₄Pt₂
MDL Number: MFCD00058713
CAS Number: 12073-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR