Dichloro(1,5-cyclooctadiene)platinum(II) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,5-Cyclooctadieneplatinum(II) Dichloride
78-0430-250MG
78-0430-1G
78-0430-5G
101442-342EA
76.43
USD
101442-342
100201-024
100201-026
Dichloro(1,5-cyclooctadiene)platinum(II) ≥99%
Dichloro(1,5-cyclooctadiene)platinum(II)
Formula:
C₈H₁₂Cl₂Pt MW: 374.17 g/mol Melting Pt: 250 °C (dec.) |
MDL Number:
MFCD00012413 CAS Number: 12080-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...