cis-Dichlorobis(diethylsulfide)platinum(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

78-0317-1G 78-0317-5G
100199-990EA 244.86 USD
100199-990 100197-612
cis-Dichlorobis(diethylsulfide)platinum(II) ≥99%
cis-Dichlorobis(diethylsulfide)platinum(II)
Formula: Pt[(C₂H₅)₂S]₂Cl₂
MW: 446.36 g/mol
MDL Number: MFCD00050943
CAS Number: 15442-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR