Bis(tri-tert-butylphosphine)platinum(0) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(tri-t-butylphosphine)platinum (0)

78-0140-1G 78-0140-250MG
100207-434EA 767.14 USD
100207-434 100197-610
Bis(tri-tert-butylphosphine)platinum(0) ≥98%
Bis(tri-tert-butylphosphine)platinum(0)
Formula: C₂₄H₅₄P₂Pt
MW: 599.73 g/mol
MDL Number: MFCD04974248
CAS Number: 60648-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR