Platinum-ruthenium/tetra-n-octylammonium chloride (∼7% Pt, ∼3.5% Ru) (w/w), colloidal

Supplier: STREM CHEMICALS INC MS
Danger

78-0062-250MG 78-0062-1G
100196-368EA 765.72 USD
100196-368 100201-252
Platinum-ruthenium/tetra-n-octylammonium chloride (∼7% Pt, ∼3.5% Ru) (w/w), colloidal
Platinum-ruthenium/tetra-n-octylammonium chloride

Nanomaterials

MDL Number: MFCD06798797
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR