Platinum/tetra-n-octylammonium chloride colloid (≥70-85% Pt), purified

Supplier: STREM CHEMICALS INC MS
Warning

78-0055-1G 78-0055-250MG
100201-250EA 2304.25 USD
100201-250 100196-366
Platinum/tetra-n-octylammonium chloride colloid (≥70-85% Pt), purified
Platinum/tetra-n-octylammonium chloride colloid

Nanomaterials

Formula: Pt/(C₈H₁₇)₄NCl
MDL Number: MFCD06798798
CAS Number: 7440-06-4
UN: 3200
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR