Tris(norbornadiene)(acetylacetonato)iridium(III) ≥98% (99.9%-Ir)
Supplier: STREM CHEMICALS INC MS
77-9700-100MG
77-9700-500MG
100197-606EA
67.94
USD
100197-606
100197-608
Tris(norbornadiene)(acetylacetonato)iridium(III) ≥98% (99.9%-Ir)
Tris(norbornadiene)(acetylacetonato)iridium(III)
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₂₆H₃₁IrO₂ Melting Pt: 189 °C |
MDL Number:
MFCD28167048 CAS Number: 41612-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...