(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I)hexafluorophosphate, CRABTREE'S CATALYST, (1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)-iridium(I) hexafluorophosphate, (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium (I) hexafluorophosphate, Crabtree's Catalyst

77-9500-250MG 77-9500-50MG
100201-742EA 247.69 USD
100201-742 100205-200
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate ≥99%
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Formula: C₃₁H₅₀F₆IrNP₂
MW: 804.90 g/mol
MDL Number: MFCD00075097
CAS Number: 64536-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR