Potassium pentachloronitrosyl iridium(III) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

77-6590-5G 77-6590-1G
100204-104EA 1917.85 USD
100204-104 100207-744
Potassium pentachloronitrosyl iridium(III) ≥99%
Potassium pentachloronitrosyl iridium(III)
MDL Number: MFCD00049790
CAS Number: 22594-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR