((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-[COD]Ir[Ph2PThrePHOX]

77-5020-100MG 77-5020-500MG
100210-092EA 165.6 USD
100210-092 100204-652
((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%
((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate

ThrePHOX

Melting Pt: 112…113 °C
MDL Number: MFCD06658140
CAS Number: 405235-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR