((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

77-5019-100MG 77-5019-500MG
101419-132EA 165.6 USD
101419-132 101419-134
((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%
((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (diphenylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate

ThrePHOX

Melting Pt: 112…113 °C
MDL Number: MFCD18253385
CAS Number: 880262-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR