((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-[COD]Ir[cy2PThrePHOX]

77-5010-100MG 77-5010-500MG
100210-090EA 165.6 USD
100210-090 100204-650
((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%
((4S,5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl]-dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate

ThrePHOX

Melting Pt: 160…161 °C
MDL Number: MFCD06658139
CAS Number: 583844-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR