((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-[COD]Ir[cy2PThrePHOX]

77-5009-500MG 77-5009-100MG
101419-130EA 607.2 USD
101419-130 101419-128
((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate ≥97%
((4R,5R)-(+)-O-[1-Benzyl-1-(5-methyl-2-phenyl-4,5-dihydrooxazol-4-yl)-2-phenylethyl] (dicyclohexylphosphinite)(1,5-COD)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenylborate

ThrePHOX

Melting Pt: 160…161 °C
MDL Number: MFCD18252665
CAS Number: 880262-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR