Carbonylhydridotris(triphenylphosphine)iridium(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hydridocarbonyltris(triphenylphosphine)iridium(I)

77-1400-5G 77-1400-1G
100208-680EA 1290.83 USD
100208-680 100208-678
Carbonylhydridotris(triphenylphosphine)iridium(I) ≥99%
Carbonylhydridotris(triphenylphosphine)iridium(I)
Formula: C₅₅H₄₆IrOP₃
MW: 1008.12 g/mol
Melting Pt: 170 °C (dec.)
MDL Number: MFCD00015525
CAS Number: 17250-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR