3-Di-i-propylphosphino-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)iridium(I) hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

77-1115-1G 77-1115-250MG
101419-126EA 636.92 USD
101419-126 101419-124
3-Di-i-propylphosphino-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)iridium(I) hexafluorophosphate ≥98%
3-Di-i-propylphosphino-2-(N,N-dimethylamino)-1H-indene(1,5-cyclooctadiene)iridium(I) hexafluorophosphate
MDL Number: MFCD26940210
CAS Number: 870077-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR