Hexachloroiridic(IV) acid hydrate ≥99.9% (Ir basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dihydrogen hexachloroiridate(IV) xhydrate (Chunks or pellets), Hydrogen iridium hexachloride, Hydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Dihydrogen hexachloroiridate(IV) hydrate

77-1090-5G 77-1090-1G
100203-818EA 1275.26 USD
100203-818 100203-816
Hexachloroiridic(IV) acid hydrate ≥99.9% (Ir basis)
Hexachloroiridic(IV) acid hydrate
Formula: H₂Cl₆Ir·xH₂O
MW: 406.93 g/mol (anhy)
MDL Number: MFCD00149906
CAS Number: 110802-84-1
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR