1,5-Cyclooctadiene(η5-indenyl)iridium(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

77-0950-250MG 77-0950-1G
101324-822EA 178.34 USD
101324-822 101324-824
1,5-Cyclooctadiene(η5-indenyl)iridium(I) ≥99%
1,5-Cyclooctadiene(η5-indenyl)iridium(I)
Formula: C₁₇H₁₉Ir
MDL Number: MFCD07782020
CAS Number: 102525-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR