1,5-Cyclooctadiene(hexafluoroacetylacetonato)iridium(I) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1,5-Cyclooctadiene)(hexafluoroacetylacetonato)iridium(I)

77-0930-500MG 77-0930-100MG
101324-820EA 309.97 USD
101324-820 101324-818
1,5-Cyclooctadiene(hexafluoroacetylacetonato)iridium(I) ≥98%
1,5-Cyclooctadiene(hexafluoroacetylacetonato)iridium(I)

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₃H₁₃F₆IrO₂
MW: 507.4500 g/mol
MDL Number: MFCD07782019
CAS Number: 34801-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR