(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate ≥99% (99.9%-Ir)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Ir(acac)(COD), (Acetylacetonato)(1,5-cyclooctadiene)iridium(I), Acetylacetonato(1,5-cyclooctadiene)iridium(I), 1,5-Cyclooctadiene(acetylacetonato)iridium(I), Ir(acac)(cod)
77-0900-500MG
77-0900-100MG
100197-600EA
277.42
USD
100197-600
100197-598
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate ≥99% (99.9%-Ir)
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₃H₁₉O₂Ir MW: 399.51 g/mol Melting Pt: 145…150 °C (dec.) |
MDL Number:
MFCD08273779 CAS Number: 12154-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...