Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chlorodihydrido[bis(2-di-i-propylphosphinoethyl)amine]iridium(III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM (III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM(III)

77-0500-250MG 77-0500-1G
101324-814EA 188.25 USD
101324-814 101324-816
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium ≥98%
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium

Pincer Ligands and Complexes

Formula: C₁₆H₄₂ClIrN₂P₂
MDL Number: MFCD07782018
CAS Number: 791629-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR