Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: [Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer

77-0400-500MG 77-0400-2G 77-0400-10G
100208-956EA 219.38 USD
100208-956 100208-958 100195-922
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥99%
Chloro(1,5-cyclooctadiene)iridium (I) dimer

Photocatalyst

Formula: C₁₆H₂₄Cl₂Ir₂
MW: 671.70 g/mol
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00012414
CAS Number: 12112-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR