Carbonylchlorobis(triphenylphosphine)iridium(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Carbonylchloro bis(triphenylphosphine)iridium(I), VASKA'S COMPLEX, Chlorocarbonylbis(triphenylphosphine)iridium(I)

77-0300-1G 77-0300-5G
100210-080EA 215.14 USD
100210-080 100210-082
Carbonylchlorobis(triphenylphosphine)iridium(I) ≥99%
Carbonylchlorobis(triphenylphosphine)iridium(I)
Formula: C₃₇H₃₀ClIrOP₂
MW: 780.26 g/mol
MDL Number: MFCD00009590
CAS Number: 14871-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR