Chlorobis(cyclooctene)iridium(I) dimer ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Chlorobis(cyclooctene)iridium(I)dimer

77-0250-1G 77-0250-250MG
101324-812EA 232.12 USD
101324-812 101324-810
Chlorobis(cyclooctene)iridium(I) dimer ≥97%
Chlorobis(cyclooctene)iridium(I) dimer
Formula: C₃₂H₅₆Cl₂Ir₂
MW: 896.134 g/mol
MDL Number: MFCD00213465
CAS Number: 12246-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR