Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(cyclooctadiene)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate

77-0225-500MG 77-0225-100MG
101442-320EA 298.65 USD
101442-320 101442-318
Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥98%
Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Formula: C₄₈H₂₈BF₂₄Ir
MDL Number: MFCD10567389
CAS Number: 666826-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR