Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate

77-0200-2G 77-0200-500MG
100201-248EA 876.12 USD
100201-248 100196-362
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥98%
Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate
Formula: C₁₆H₂₄BF₄Ir
MW: 495.383 g/mol
MDL Number: MFCD00079664
CAS Number: 35138-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR