Ammonium hexachloroiridate(III) hydrate (∼39% Ir)

Supplier: STREM CHEMICALS INC MS
Warning

77-0019-1G 77-0019-5G
100210-076EA 270.34 USD
100210-076 100210-078
Ammonium hexachloroiridate(III) hydrate (∼39% Ir)
Ammonium hexachloroiridate(III) hydrate
Formula: (NH₄)₃IrCl₆·xH₂O
MDL Number: MFCD00003393
CAS Number: 15752-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR