Bis(cyclopentadienyl)osmium ≥99% (99.9%-Os)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Osmocene

76-0150-1G 76-0150-250MG
100205-984EA 903.02 USD
100205-984 100200-300
Bis(cyclopentadienyl)osmium ≥99% (99.9%-Os)
Bis(cyclopentadienyl)osmium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Os
MW: 320.42 g/mol
Melting Pt: 226…228 °C
MDL Number: MFCD00058734
CAS Number: 1273-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR