i-Propylcyclopentadienylrhenium tricarbonyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

75-2410-250MG 75-2410-1G
100197-590EA 778.46 USD
100197-590 100211-164
i-Propylcyclopentadienylrhenium tricarbonyl ≥97%
i-Propylcyclopentadienylrhenium tricarbonyl

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD00144499
CAS Number: 126250-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR