Pentamethylcyclopentadienylrhenium tricarbonyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

75-2400-2G 75-2400-500MG
100197-168EA 1007.75 USD
100197-168 100210-832
Pentamethylcyclopentadienylrhenium tricarbonyl ≥98%
Pentamethylcyclopentadienylrhenium tricarbonyl

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Melting Pt: 149…151 °C
MDL Number: MFCD00058888
CAS Number: 12130-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR