Iododioxobis(triphenylphosphine)rhenium(V) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

75-2345-1G 75-2345-5G
100203-286EA 154.28 USD
100203-286 100203-288
Iododioxobis(triphenylphosphine)rhenium(V) ≥98%
Iododioxobis(triphenylphosphine)rhenium(V)
MDL Number: MFCD00151571
CAS Number: 23032-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR