Cyclopentadienylrhenium tricarbonyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

75-2300-5G 75-2300-1G
100201-734EA 1579.57 USD
100201-734 100201-732
Cyclopentadienylrhenium tricarbonyl ≥99%
Cyclopentadienylrhenium tricarbonyl

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₈H₅O₃Re
MDL Number: MFCD00058885
CAS Number: 12079-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR