Rhenium ≥99.97%, wire

Supplier: STREM CHEMICALS INC MS
Warning

75-0090-10CM 75-0090-50CM
100198-250EA 140.12 USD
100198-250 100208-054
Rhenium ≥99.97%, wire
Rhenium
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5900 °C
Melting Pt: 3180 °C
Density: 21,04
MDL Number: MFCD00011195
CAS Number: 7440-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR