Rhenium ≥99.99%, pellets

Supplier: STREM CHEMICALS INC MS
Warning

75-0075-2G 75-0075-10G 75-0075-50G
100204-102EA 209.48 USD
100204-102 100201-726 100204-646
Rhenium ≥99.99%, pellets
Rhenium
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5900 °C
Melting Pt: 3180 °C
Density: 21.04
MDL Number: MFCD00011195
CAS Number: 7440-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR