Tris(t-butoxy)(2,2-dimethylpropylidyne)tungsten(VI) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

74-1800-100MG 74-1800-500MG
100210-058EA 523.69 USD
100210-058 100210-664
Tris(t-butoxy)(2,2-dimethylpropylidyne)tungsten(VI) ≥98%
Tris(t-butoxy)(2,2-dimethylpropylidyne)tungsten(VI)
MDL Number: MFCD06798309
CAS Number: 78234-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR