(Mesitylene)tungsten tricarbonyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

74-1350-1G 74-1350-5G
100208-050EA 247.69 USD
100208-050 100208-052
(Mesitylene)tungsten tricarbonyl ≥98%
(Mesitylene)tungsten tricarbonyl

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₂H₁₂WO₃
MW: 388.065 g/mol
Melting Pt: 165 °C (dec.)
MDL Number: MFCD00015319
CAS Number: 12129-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR