Tantalum(V) tetraethoxide 2,4-pentanedionate ≥99.99% (Ta basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tantalum(V) (tetraethoxy)(acetylacetonate)

73-5000-10G 73-5000-50G
100203-280EA 280.25 USD
100203-280 100203-282
Tantalum(V) tetraethoxide 2,4-pentanedionate ≥99.99% (Ta basis), PURATREM
Tantalum(V) tetraethoxide 2,4-pentanedionate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₃H₂₇O₆Ta
MW: 460.298 g/mol
Boiling Pt: 95 °C (0.5 mmHg)
Melting Pt: 45 °C
Density: 1.5
MDL Number: MFCD00145349
CAS Number: 20219-33-4
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR