i-Propylcyclopentadienylhafnium trichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

72-7000-1G 72-7000-5G
100207-430EA 206.65 USD
100207-430 100196-884
i-Propylcyclopentadienylhafnium trichloride ≥98%
i-Propylcyclopentadienylhafnium trichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00144498
CAS Number: 329736-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR