(Pentamethylcyclopentadienyl)hafnium trichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Pentamethylcyclopentadienylhafnium(IV) trichloride

72-6000-5G 72-6000-1G
100208-048EA 564.74 USD
100208-048 100208-046
(Pentamethylcyclopentadienyl)hafnium trichloride ≥98%
(Pentamethylcyclopentadienyl)hafnium trichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₅Cl₃Hf
MW: 420.07 g/mol
MDL Number: MFCD00070461
CAS Number: 75181-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR