Hafnium(IV) isopropoxide isopropanol adduct ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hafnium(IV) i-propoxide monoisopropylate

72-5950-25G 72-5950-5G
100210-048EA 866.22 USD
100210-048 100210-050
Hafnium(IV) isopropoxide isopropanol adduct ≥99%
Hafnium(IV) isopropoxide isopropanol adduct

Alkoxide

Formula: C₁₂H₂₈HfO₄·C₃H₈O
MW: 474.94 g/mol
MDL Number: MFCD00070460
CAS Number: 2171-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR