Hafnium(IV) ethanolate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hafnium ethoxide, Hafnium(IV) ethoxide

72-5900-25G 72-5900-5G
100207-736EA 850.65 USD
100207-736 100207-738
Hafnium(IV) ethanolate ≥99%
Hafnium (IV) ethanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₈H₂₀HfO₄
MW: 358.732 g/mol
MDL Number: MFCD00070459
CAS Number: 13428-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR