Hafnium(IV) acetylacetonate ≥96%

Supplier: STREM CHEMICALS INC MS

Synonyms: Hafnium(IV) 2,4-pentanedionate

72-2002-25G 72-2002-5G 72-2002-1G
100208-950EA 651.08 USD
100208-950 100197-584 100207-734
Hafnium(IV) acetylacetonate ≥96%
Hafnium (IV) acetylacetonate
Formula: C₂₀H₂₈O₈Hf
MDL Number: MFCD00044997
CAS Number: 17475-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR