Bis(i-propylcyclopentadienyl)hafnium dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

72-2000-5G 72-2000-1G
100205-980EA 1111.08 USD
100205-980 100210-660
Bis(i-propylcyclopentadienyl)hafnium dichloride ≥98%
Bis(i-propylcyclopentadienyl)hafnium dichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD01863548
CAS Number: 66349-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR