Dimethylbis(t-butylcyclopentadienyl)hafnium(IV) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

72-1900-5G 72-1900-1G 72-1900-25G
100205-978EA 1201.66 USD
100205-978 100210-658 100205-976
Dimethylbis(t-butylcyclopentadienyl)hafnium(IV) ≥98%
Dimethylbis(t-butylcyclopentadienyl)hafnium(IV)

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD01073804
CAS Number: 68193-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR