Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

72-1500-5G 72-1500-1G
100199-974EA 608.62 USD
100199-974 100203-264
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride ≥98%
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀Cl₂Hf
MDL Number: MFCD00058887
CAS Number: 85959-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR