Bis(ethylcyclopentadienyl)hafnium dichloride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
72-1250-1G
72-1250-5G
100207-428EA
246.28
USD
100207-428
100196-962
Bis(ethylcyclopentadienyl)hafnium dichloride ≥98%
Bis(ethylcyclopentadienyl)hafnium dichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₄H₁₈Cl₂Hf |
MDL Number:
MFCD0014376 CAS Number: 78205-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...